Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$188.28 - $2109.88
Specifications
| Antigen | Erythropoietin R |
|---|---|
| Clone | 38409 |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
| Erythropoietin R | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| EPOR | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 38409 | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P19235 | |
| 2057 | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Erythropoietin R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Erythropoietin R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title