Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FATP5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | FATP5/SLC27A5 |
---|---|
Clone | 390917 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
FATP5/SLC27A5 Monoclonal specifically detects FATP5/SLC27A5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FATP5/SLC27A5 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q9Y2P5 | |
10998 | |
HEK293 human embryonic kidney cell line transfected with human FATP5 Met1-Leu690 Accession # Q9Y2P5 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FATP5. Stains human FATP5 transfectants but not irrelevant transfectants. |
390917 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACSB, ACSBBACS, ACSVL6, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, SLC27A5, solute carrier family 27 (fatty acid transporter), member 5, VLACSR, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related | |
SLC27A5 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title