Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB238NA
Change view
Click to view available options
Gene Accession No.:
P10767
P10767.4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gene Accession No.
AB238NA P10767.4
AF238 P10767
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AB238NA Supplier R&D Systems Supplier No. AB238NA
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

FGF-6 Polyclonal specifically detects FGF-6 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen FGF-6
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 15-25 ug/mL
Gene Accession No. P10767.4
Gene Alias FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2
Gene Symbols FGF6
Host Species Goat
Immunogen E. coli-derived recombinant human FGF-6 Gly41-Ile208 Accession # P10767.4
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2251
Test Specificity Detects human FGF-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 17% cross-reactivity with recombinant human (rh) FGF-4 is observed and less then 1% cross-reactivity with rhFGF acidic, rhFGF basic, and rhFGF-5 is observed. Neutralizes the biological activity of rhFGF-6, but will not neutralize the biological activity of rhFGF acidic, rhFGF basic, rhFGF-4, or rhFGF-5.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.