Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGF-6 Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody has been used in 2 publications

$526.50 - $549.50

Specifications

Antigen FGF-6
Concentration LYOPH
Applications Western Blot, Neutralization
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Gene Accession No. Price Quantity & Availability  
Catalog Number Mfr. No. Gene Accession No. Price Quantity & Availability  
AF238
GREENER_CHOICE
View Documents
R&D Systems
AF238
P10767
Each for $549.50
Only null left
Add to Cart
 
AB238NA
GREENER_CHOICE
View Documents
R&D Systems
AB238NA
P10767.4
Each for $526.50
Only null left
Add to Cart
 
Description

Description

FGF-6 Polyclonal specifically detects FGF-6 in Human samples. It is validated for Western Blot, Neutralization.
Specifications

Specifications

FGF-6
Western Blot, Neutralization
Unconjugated
RUO
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2
FGF6
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Polyclonal
Goat
Human
2251
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.