Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB684500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB684500 500 μg
MAB684SP 25 μg
MAB684100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB684500 Supplier R&D Systems Supplier No. MAB684500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen FGFR2
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 98725
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 1-5 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Gene Symbols FGFR2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2263
Test Specificity Detects alpha isoforms (3 Ig-like domains) and beta isoforms (2 Ig-like domains) of human FGF R2 in direct ELISAs and Western blots. In direct ELISAs, this antibody reacts with all isoforms of rhFGF R2 but shows approximately a 4-fold preference for (IIIc) isoforms. Approximately 100% cross-reactivity with recombinant mouse FGF R2, 15% cross-reactivity with recombinant human (rh) FGF RI ( beta isoforms), and no cross-reactivity with rhFGF RI ( alpha isoforms), rhFGF R3, or rhFGF R4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.