Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB7661100
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Neutralization.Specifications
FGFR3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGFR3 | |
Mouse myeloma cell line NS0-derived recombinant human FGF R3 alpha (IIIc) | |
100 μg | |
Primary | |
Detects the IIIb and IIIc isoforms of human FGF R3. It does not cross-react with mouse FGF R3 or with any isoforms of recombinant human (rh) FGF R1 or rhFGF R2. | |
Human | |
Supernatant |
Western Blot, Neutralization | |
136318 | |
Western Blot 1 ug/mL, Neutralization 2.5-15 ng/mL | |
CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2261 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction