Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FGFR3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5317149
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7661SP 25 μg
MAB7661100 100 μg
MAB7661500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB7661SP Supplier R&D Systems Supplier No. MAB7661SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen FGFR3
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 136318
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 2.5-15 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
Gene Symbols FGFR3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FGF R3 alpha (IIIc)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2261
Test Specificity Detects the IIIb and IIIc isoforms of human FGF R3. It does not cross-react with mouse FGF R3 or with any isoforms of recombinant human (rh) FGF R1 or rhFGF R2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.