Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fgr Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB32071
This item is not returnable.
View return policy
Description
Fgr Monoclonal specifically detects Fgr in Human samples. It is validated for Immunocytochemistry.Specifications
Fgr | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P09769 | |
FGR | |
E. coli-derived recombinant human Fgr Met1-Phe82 Accession # P09769 | |
100 μg | |
Primary | |
Detects human Fgr in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
349331 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-fgr, c-src2, c-src-2 proto-oncogene, EC 2.7.10, EC 2.7.10.2, FLJ43153, Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog, MGC75096, p55c-fgr, p55-c-fgr protein, p55-Fgr, p58c-fgr, Proto-oncogene c-Fgr, proto-oncogene tyrosine-protein kinase FGR, Src2, SRC2c-fgr protooncogene, tyrosine-protein kinase Fgr | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2268 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction