Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fgr Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Fgr |
---|---|
Clone | 349331 |
Concentration | LYOPH |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB32071SP
![]() |
R&D Systems
MAB32071SP |
25 μg |
Each for $200.50
|
|
|||||
MAB32071
![]() |
R&D Systems
MAB32071 |
100 μg |
Each for $491.00
|
|
|||||
Description
Fgr Monoclonal specifically detects Fgr in Human samples. It is validated for Immunocytochemistry.Specifications
Fgr | |
LYOPH | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P09769 | |
2268 | |
E. coli-derived recombinant human Fgr Met1-Phe82 Accession # P09769 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Fgr in direct ELISAs. |
349331 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-fgr, c-src2, c-src-2 proto-oncogene, EC 2.7.10, EC 2.7.10.2, FLJ43153, Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog, MGC75096, p55c-fgr, p55-c-fgr protein, p55-Fgr, p58c-fgr, Proto-oncogene c-Fgr, proto-oncogene tyrosine-protein kinase FGR, Src2, SRC2c-fgr protooncogene, tyrosine-protein kinase Fgr | |
FGR | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title