Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibrinogen A Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Fibrinopeptide A |
---|---|
Clone | 949330 |
Dilution | Western Blot 1 μg;/mL, Immunohistochemistry 5-25 μg;/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9898SP
![]() |
R&D Systems
MAB9898SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9898100
![]() |
R&D Systems
MAB9898100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Fibrinopeptide A Monoclonal specifically detects Fibrinopeptide A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Fibrinopeptide A | |
Western Blot 1 μg;/mL, Immunohistochemistry 5-25 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
P02671 | |
2243.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
949330 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FGA, Fib2, fibrinogen alpha chain, Fibrinogen alpha subunit, Fibrinogen Type I, fibrinogen, A alpha polypeptide, FPA, MGC119422, MGC119423, MGC119425 | |
Synthetic peptide containing human Fibrinogen A Peptide, Accession No. P02671 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Fibrinogen A in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title