Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fibrinogen Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Fibrinogen |
---|---|
Clone | 949332 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9404SP
![]() |
R&D Systems
MAB9404SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9404100
![]() |
R&D Systems
MAB9404100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Fibrinopeptide B Monoclonal specifically detects Fibrinopeptide B in Human samples. It is validated for Immunocytochemistry.Specifications
Fibrinogen | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P02671 | |
2243 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
949332 | |
Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Factor I, Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
Human serum-derived Fibrinogen, Accession # P02671 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Fibrinogen in direct ELISAs. In Western blots, does not detect Fibrinogen A. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title