Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ficolin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4209SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4209SP 25 μg
AF4209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4209SP Supplier R&D Systems Supplier No. AF4209SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Ficolin-1 Polyclonal specifically detects Ficolin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ficolin-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00602
Gene Alias Collagen/fibrinogen domain-containing protein 1, FCN1, Fcna, FCNM, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin1, Ficolin-A, ficolin-alpha, M-ficolin
Gene Symbols FCN1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Ficolin-1 Gln28-Ala326 Accession # O00602
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2219
Test Specificity Detects human Ficolin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) Ficolin-2 is observed and less than 1% cross-reactivity with rhFicolin-3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.