Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human Ficolin-1 Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Sheep Polyclonal Antibody
$200.50 - $514.50
Specifications
| Antigen | Ficolin-1 | 
|---|---|
| Concentration | LYOPH | 
| Dilution | Western Blot 0.1 ug/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
Description
Ficolin-1 Polyclonal specifically detects Ficolin-1 in Human samples. It is validated for Western Blot.Specifications
| Ficolin-1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| Collagen/fibrinogen domain-containing protein 1, FCN1, Fcna, FCNM, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin1, Ficolin-A, ficolin-alpha, M-ficolin | |
| FCN1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O00602 | |
| 2219 | |
| Mouse myeloma cell line NS0-derived recombinant human Ficolin-1 Gln28-Ala326 Accession # O00602 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Ficolin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) Ficolin-2 is observed and less than 1% cross-reactivity with rhFicolin-3 is observed. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title