Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Follistatin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
$200.50 - $2020.00
Specifications
Antigen | Follistatin |
---|---|
Clone | 85918 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Follistatin | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P19883 | |
10468 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Follistatin Gly30-Asp329 Accession # P19883 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Follistatin in direct ELISAs and Western blots. It recognizes the 288 aa, 300 aa, and 315 aa isoforms of human Follistatin. In direct ELISAs and Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse Follistatin. |
85918 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
follistatin, follistatin isoform FST317, FS, FSActivin-binding protein, FST | |
FST | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title