Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1153SP
This item is not returnable.
View return policy
Description
Galectin-2 Polyclonal specifically detects Galectin-2 in Human samples. It is validated for Western Blot.Specifications
Galectin-2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
Goat | |
Affinity Purified | |
RUO | |
3957 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
P05162 | |
LGALS2 | |
E. coli-derived recombinant human Galectin-2 (R&D Systems, Catalog # 1153-GA) Met1-Glu132 Accession # P05162 | |
25 μg | |
Primary | |
Detects human Galectin-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-4, rhGalectin-7, and rhGalectin-8 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction