Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Galectin-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Galectin-2 Polyclonal specifically detects Galectin-2 in Human samples. It is validated for Western Blot.Specifications
Galectin-2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
LGALS2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P05162 | |
3957 | |
E. coli-derived recombinant human Galectin-2 (R&D Systems, Catalog # 1153-GA) Met1-Glu132 Accession # P05162 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Galectin-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-4, rhGalectin-7, and rhGalectin-8 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title