Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GASP-1/WFIKKN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | GASP-1/WFIKKN2 |
---|---|
Clone | 288518 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB20701SP
![]() |
R&D Systems
MAB20701SP |
25 μg |
Each for $200.50
|
|
|||||
MAB20701
![]() |
R&D Systems
MAB20701 |
100 μg |
Each for $491.00
|
|
|||||
Description
GASP-1/WFIKKN2 Monoclonal specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for Immunohistochemistry.Specifications
GASP-1/WFIKKN2 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q8TEU8 | |
124857 | |
Mouse myeloma cell line NS0-derived recombinant human GASP-1/WFIKKN2 Leu35-His576 Accession # Q8TEU8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GASP-1/WFIKKN2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-2 is observed. |
288518 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN2, WFIKKN-related protein, WFIKKNRP, WFIKKNRPmultivalent protease inhibitor protein | |
WFIKKN2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title