Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GATA-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$188.28 - $520.12
Specifications
| Antigen | GATA-1 |
|---|---|
| Clone | 234732 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
Description
GATA-1 Monoclonal specifically detects GATA-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| GATA-1 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P15976 | |
| 2623 | |
| E. coli-derived recombinant human GATA-1 Met1-Ser413 Accession # P15976 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GATA-1 in direct ELISAs. In Western blots, this antibody does not cross-react with rhGATA-2 or rhGATA-6. |
| 234732 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA1, GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT | |
| GATA1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title