Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-5/BMP-14 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$160.80 - $474.00
Specifications
Antigen | GDF-5/BMP-14 |
---|---|
Clone | 1065032 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8340SP
![]() |
R&D Systems
MAB8340SP |
25 μg |
Each for $160.80
|
|
|||||
MAB8340100
![]() |
R&D Systems
MAB8340100 |
100 μg |
Each for $474.00
|
|
|||||
Description
GDF-5/BMP-14 Monoclonal antibody specifically detects GDF-5/BMP-14 in Human samples. It is validated for ImmunocytochemistrySpecifications
GDF-5/BMP-14 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
E. coli-derived recombinant human GDF-5, Ala382-Arg501, Accession # P43026 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1065032 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P43026 | |
8200 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title