Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDPD5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | GDPD5 |
---|---|
Clone | 506020 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10524SP
![]() |
R&D Systems
MAB10524SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10524100
![]() |
R&D Systems
MAB10524100 |
100 μg |
Each for $432.50
|
|
|||||
Description
GDPD5 Monoclonal specifically detects GDPD5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
GDPD5 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
EC 3.1, GDE2, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5, PP1665 | |
Human embryonic kidney cell, HEK293-derived transfected with GDPD5, Accession # Q8WTR4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
506020 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q8WTR4 | |
81544 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title