Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human gp130 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $2020.00

Specifications

Antigen gp130/CD130
Clone 28105
Dilution Immunohistochemistry 5-25 ug/mL, Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Immunohistochemistry, Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB628SP
GREENER_CHOICE
View Documents
R&D Systems
MAB628SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB628100
GREENER_CHOICE
View Documents
R&D Systems
MAB628100
100 μg
Each for $538.00
Only null left
Add to Cart
 
MAB628500
GREENER_CHOICE
View Documents
R&D Systems
MAB628500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

gp130/CD130
Immunohistochemistry 5-25 ug/mL, Neutralization 0.02-0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P40189
3572
Sf 21-derived recombinant human gp130 extracellular domain Leu24-Glu619 (Glu619-Asp, predicted) Accession # P40189
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human gp130 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) CNTF, rhIL-6, rhIL-6 R, recombinant mouse (rm) IL-6, rhIL-11, rhLIF, rhLIF R, rmLIF, or rhOSM is observed. This antibody blocks the human gp130-mediated bioactivities induced by IL-6, IL-11, LIF, OSM, and CNTF.
28105
Immunohistochemistry, Neutralization, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
IL6ST
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.