Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Growth Hormone Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.5 mg
1 mg
Description
Recombinant bioactive protein containing 192 amino acids for Human Growth Hormone The Recombinant Human Growth Hormone Protein is derived from E. coli. The Recombinant Human Growth Hormone Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Accession Number | P01241 |
| Formulation | Lyophilized |
| Gene ID (Entrez) | 2688 |
| Name | Growth Hormone Protein |
| Purification Method | SDS-PAGE |
| Quantity | 0.5 mg |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Reconstitution | Reconstitute in 10 mM Sodium phosphate buffer (pH 8.0) or 10 mM NH4HCO3 (pH 8.0). The solution can then be further diluted to other aqueous solutions. When diluting to below 10 ug/ml, the diluted solution should contain carrier protein (BSA or others) to |
| Purity or Quality Grade | >98%, by SDS-PAGE |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction