Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HIN-1/SCGB3A1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
| Antigen | HIN-1/SCGB3A1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HIN-1/SCGB3A1 Polyclonal specifically detects HIN-1/SCGB3A1 in Human samples. It is validated for Western Blot.Specifications
| HIN-1/SCGB3A1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867 | |
| SCGB3A1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q96QR1 | |
| 92304 | |
| E. coli-derived recombinant human HIN-1/SCGB3A1 Phe21-Gly104 Accession # Q96QR1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human HIN-1/SCGB3A1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title