Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IBSP/Sialoprotein II Antibody, R&D Systems™
Goat Polyclonal Antibody
Specifications
Antigen | IBSP/Sialoprotein II |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IBSP/Sialoprotein II Polyclonal specifically detects IBSP/Sialoprotein II in Human samples. It is validated for Western Blot.Specifications
IBSP/Sialoprotein II | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
BNSP, Bone sialoprotein, Bone sialoprotein 2, BSP, BSP 2, BSP II, BSP2, BSPII, BSP-II, Cell binding sialoprotein, IBSP, Integrin binding sialoprotein, SP II, SPII, SP-II | |
IBSP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAC95490 | |
3381 | |
Chinese hamster ovary cell line CHO-derived recombinant human IBSP/Sialoprotein II Phe17-Gln317 Accession # AAC95490 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IBSP/Sialoprotein II in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title