Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
$472.87
Specifications
| Antigen | ICAM-3/CD50 |
|---|---|
| Clone | Cal 3.10 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready.Specifications
| ICAM-3/CD50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 3385 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant |
| Cal 3.10 | |
| Unconjugated | |
| Mouse | |
| Human | |
| Recombinant human ICAM-3/CD50 Fc Chimera | |
| Primary | |
| Detects human ICAM-3/CD50 in Western blots. Does not cross-react with recombinant human (rh) ICAM-1 or rhVCAM-1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title