Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$164.60 - $544.24
Specifications
| Antigen | ICAM-3/CD50 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Polyclonal |
Description
ICAM-3/CD50 Polyclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| ICAM-3/CD50 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Polyclonal | |
| Sheep | |
| Human | |
| 3385 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ICAM-3/CD50 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ICAM-3/CD50 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) ICAM-1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) ICAM-1, recombinant rat ICAM-1, rhICAM-2, rhVCAM-1, and rmVCAM-1 is observed. |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| ICAM3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title