Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7083288
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8142SP 25 μg
MAB8142100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8142SP Supplier R&D Systems Supplier No. MAB8142SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen IFN-beta
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2036A
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 uL/10^6 cells, Immunocytochemistry 1-25 ug/mL
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P01574
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta Met1-Asn187 Accession # P01574
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Detects human IFN-beta in direct ELISAs.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.