Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB91472SP
This item is not returnable.
View return policy
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for Immunocytochemistry.Specifications
IFN-epsilon | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020 | |
E. coli-derived recombinant human IFN-epsilon, Leu22-Arg208, Accession No. Q86WN2 | |
25 μg | |
338376.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG1 |
Immunocytochemistry | |
995728 | |
Immunocytochemistry 8-25 μg;/mL | |
Q86WN2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human IFN-epsilon in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction