Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-epsilon Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $502.50

Specifications

Antigen IFN-epsilon
Clone 995728
Dilution Immunocytochemistry 8-25 μg;/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB91472SP
GREENER_CHOICE
View Documents
R&D Systems
MAB91472SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MB91472100
GREENER_CHOICE
View Documents
R&D Systems
MAB91472100
100 μg
Each for $502.50
Only null left
Add to Cart
 
Description

Description

IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

IFN-epsilon
Immunocytochemistry 8-25 μg;/mL
Monoclonal
Supernatant
Human
Q86WN2
338376.0
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
995728
Immunocytochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020
E. coli-derived recombinant human IFN-epsilon, Leu22-Arg208, Accession No. Q86WN2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IFN-epsilon in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.