Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ig Lambda Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
| Antigen | Human lambda light chain |
|---|---|
| Clone | 1000040 |
| Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB100491SP
![]() |
R&D Systems
MAB100491SP |
25 μg |
Each for $200.50
|
|
|||||
MB100491100
![]() |
R&D Systems
MAB100491100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Human lambda light chain Monoclonal specifically detects Human lambda light chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
| Human lambda light chain | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Ig Lamda light chain, | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Human Ig Lambda Light Chain in direct ELISAs. |
| 1000040 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| Bence Jones protein, BJP, Constant region of lambda light chains, HCAK 1, HCAK1, Ig kappa chain C region, Ig lambda chain C regions, IGKC, IGLC, IGLC 1, IGLC 2, IGLC 3, IGLC1, IGLC2, IGLC3, Immunoglobulin kappa constant, Immunoglobulin kappa constant region, Immunoglobulin kappa light chain, Immunoglobulin lambda constant 1, Kappa 1 immunoglobulin light chain, Km, Mcg marker, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809, Paraprotein | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title