Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgA Antibody (BFG-267), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB310120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310120 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310120 Supplier Novus Biologicals Supplier No. NBP142270AF750

Mouse Monoclonal Antibody

Human IgA Monoclonal antibody specifically detects Human IgA in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Human IgA
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone BFG-267
Conjugate Alexa Fluor 750
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CD79A;IGA;Igalpha;Ig-alpha;Ly54;Ly-54;MB1;MB-1
Host Species Mouse
Immunogen Purified alpha-chain (Fc) of human IgA
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.