Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgA Antibody (BFG-267), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP142270FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142270FR 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142270FR Supplier Novus Biologicals Supplier No. NBP142270FR

Mouse Monoclonal Antibody

Human IgA Monoclonal specifically detects Human IgA in Human samples. It is validated for ELISA.

Specifications

Antigen Human IgA
Applications ELISA
Classification Monoclonal
Clone BFG-267
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Purified alpha-chain (Fc) of human IgA
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody reacts with human IgA, alpha-chain. No cross reaction with human plasma proteins other then human IgA. No cross reaction with purified human IgE, IgG and IgM. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.