Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgA Antibody, Novus Biologicals™
SDP

Catalog No. NBP142092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142092 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142092 Supplier Novus Biologicals Supplier No. NBP142092

Goat Polyclonal Antibody

Human IgA Polyclonal specifically detects Human IgA in Human samples. It is validated for ELISA.

Specifications

Antigen Human IgA
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation 50% glycerol, 0.1 M NaCl, 0.01 M sodium phosphate at pH 7.5 with No Preservative
Host Species Goat
Immunogen The antibody reacts with the Human IgA antigen.
Purification Method IgG purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Antibodies were raised in goat to human IgA. From the crude polyclonal the cross reactive antibodies were extracted by incubation with Sepharose-bound human IgG and IgM. Specific antibodies were absorpted by incubation with Sepha¡rose-bound human IgA. Specific antibodies were eluted by acidic buffer at pH 2.5 followed by neutralisation and dialysis. After repeated binding with immobilized human IgA a minimum of 65% protein bound.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.