Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IGFBP-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $667.00
Specifications
Antigen | IGFBP-6 |
---|---|
Clone | 110211 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IGFBP-6 Monoclonal specifically detects IGFBP-6 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGFBP-6 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6 | |
IGFBP6 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
110211 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
P24592 | |
3489 | |
Mouse myeloma cell line NS0-derived recombinant human IGFBP-6 Ala25-Gly240 Accession # P24592 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IGFBP-6 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IGFBP-1, -2, -3, -4, -5, rmIGFBP-3, -5, or -6. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title