Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgM Antibody (3H10G5F2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP151525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151525 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151525 Supplier Novus Biologicals Supplier No. NBP151525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human IgM Monoclonal specifically detects Human IgM in Human samples. It is validated for ELISA.

Specifications

Antigen Human IgM
Applications ELISA
Classification Monoclonal
Clone 3H10G5F2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1: 10000
Gene Alias AGM1, MU, VH
Host Species Mouse
Immunogen ChromPure Human IgM (myeloma), whole molecule.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.