Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgM Antibody (ICL-931), HRP, Novus Biologicals™
SDP

Catalog No. NBP142268H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142268H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142268H Supplier Novus Biologicals Supplier No. NBP142268H

Mouse Monoclonal Antibody

Human IgM Monoclonal specifically detects Human IgM in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen Human IgM
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Dot Blot
Classification Monoclonal
Clone ICL-931
Conjugate HRP
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Dot Blot
Formulation PBS with No Preservative
Gene Alias AGM1, MU, VH
Host Species Mouse
Immunogen Purified human IgM
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This monoclonal antibody reacts with human IgM, mu-chain. No cross reaction with human plasma proteins other then human IgM. No cross reaction with purified human IgA, IgE and IgG. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.