Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-12/IL-35 p35 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15701SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15701SP 25 μg
MAB15701100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB15701SP Supplier R&D Systems Supplier No. MAB15701SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.

Specifications

Antigen IL-12/IL-35 p35
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2038C
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P29460 (p40) & P29459 (P35)
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35 Ile23-Ser328 of p40, Arg23-Ser219 of p35 Accession # P29460 (p40) & P29459 (P35)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human IL-12/IL-35 p35 in direct ELISAs. Detects the IL-12/IL-35 p35 subunit either as part of the IL-12 p40/p35 heterodimer or as the IL-12/IL-35 p35 monomer. This antibody does not detect the IL-12/IL-23 p40 subunit.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.