Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IL-17/IL-17A ELISA Kit (Colorimetric)
SDP

Catalog No. NBP192657 Shop All Novus Biologicals Products
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Mouse IL-17/IL-17A ELISA Kit (Colorimetric) measures mouse IL-17/IL-17A in cell culture supernatant, serum.

Specifications

Assay Range 7.8 - 500pg/mL
Assay Sensitivity 1.6pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3605
Gene Symbol IL17A
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-17, Biotin-Conjugate anti-mouse IL-17 monoclonal antibody, Streptavidin-HRP, Mouse IL-17 Standard lyophilized, 1ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8
Target IL-17/IL-17A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.