Learn More
Human IL-17RA/IL-17R (Research Grade Brodalumab Biosimilar) Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IL-17RA/IL-17R |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | Hu129 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Alias | CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A |
| Host Species | Human |
| Immunogen | Human Il-17 Receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.