Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-18/IL-1F4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB91244100
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA.Specifications
IL-18/IL-1F4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
E. coli-derived recombinant human IL-18/IL-1F4 Tyr37-Asp193 Accession # Q14116 | |
100 μg | |
3606 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
ELISA | |
1072H | |
ELISA | |
Q14116 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human IL-18/IL-1F4 in direct ELISAs. In a sandwich immunoassay, with Goat Anti-Human IL-18 BPa Antigen Affinity Purified Polyclonal Antibody (Catalog # AF119 ) the human IL-18/IL-18 BPa complex is detected. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction