Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-18/IL-1F4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | IL-18/IL-1F4 |
---|---|
Clone | 1072H |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB91244SP
![]() |
R&D Systems
MAB91244SP |
25 μg |
Each for $214.00
|
|
|||||
MAB91244100
![]() |
R&D Systems
MAB91244100 |
100 μg |
Each for $526.00
|
|
|||||
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA.Specifications
IL-18/IL-1F4 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q14116 | |
3606 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1072H | |
ELISA | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
E. coli-derived recombinant human IL-18/IL-1F4 Tyr37-Asp193 Accession # Q14116 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-18/IL-1F4 in direct ELISAs. In a sandwich immunoassay, with Goat Anti-Human IL-18 BPa Antigen Affinity Purified Polyclonal Antibody (Catalog # AF119 ) the human IL-18/IL-18 BPa complex is detected. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title