Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-18 R alpha/IL-1 R5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5316946
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB840SP 25 μg
MAB840100 100 μg
MAB840500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB840SP Supplier R&D Systems Supplier No. MAB840SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen IL-18R alpha/IL-1 R5
Applications Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 70625
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.03-0.1 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13478
Gene Symbols IL18R1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Test Specificity Detects Recombinant Human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.