Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB7821500
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Neutralization.Specifications
IL-22 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
50616 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Neutralization | |
142928 | |
Neutralization 1-5 ug/mL | |
Q9GZX6 | |
IL22 | |
E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
500 μg | |
Primary | |
Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction