Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $2070.00
Specifications
Antigen | IL-22 |
---|---|
Clone | 142928 |
Dilution | Neutralization 1-5 ug/mL |
Applications | Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7821SP
![]() |
R&D Systems
MAB7821SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7821100
![]() |
R&D Systems
MAB7821100 |
100 μg |
Each for $573.00
|
|
|||||
MAB7821500
![]() |
R&D Systems
MAB7821500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Neutralization.Specifications
IL-22 | |
Neutralization 1-5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
IL22 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
142928 | |
Neutralization | |
Unconjugated | |
Mouse | |
Human | |
Q9GZX6 | |
50616 | |
E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title