Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5379964
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB603SP 25 μg
MAB603100 100 μg
MAB603500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB603SP Supplier R&D Systems Supplier No. MAB603SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-3 Monoclonal specifically detects IL-3 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-3
Applications Neutralization, ELISA
Classification Monoclonal
Clone 4815
Conjugate Unconjugated
Dilution Neutralization 0.1-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08700
Gene Alias Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Gene Symbols IL3
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-3 Ala20-Phe152 Accession # P08700
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3562
Test Specificity Detects human IL-3 in ELISAs and Western blots. In sandwich ELISAs, no significant cross-reactivity or interference with recombinant human (rh) IL-1 alpha , rhIL-1 beta , recombinant mouse (rm) IL-1 beta , rhIL-2, rhIL-3 R alpha , rhIL-3 R beta , rmIL-3, rhIL-4, rmIL-4, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, bovine FGF acidic, bovine FGF basic, rhG-CSF, rhGM-CSF, rmGM-CSF, rhLIF, hPDGF, porcine PDGF, human TGF-beta 1, porcine TGF-beta 1, rhTGF-beta 1, porcine TGF-beta 2, porcine TGF-beta 1.2, rhTNF-alpha , or rhTNF-beta was observed. Does not neutralize the biological activity of rmIL-3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.