Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-31RA Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50
Specifications
Antigen | IL-31RA |
---|---|
Clone | 313308 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2769-SP
![]() |
R&D Systems
MAB2769SP |
25 μg |
Each of 1 for $200.50
|
|
|||||
Description
IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.Specifications
IL-31RA | |
Flow Cytometry | |
Unconjugated | |
Rat | |
NP_620586 | |
133396 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-31 RA in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 RA is observed. |
313308 | |
Monoclonal | |
Supernatant | |
Human | |
class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title