Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-36 gamma/IL-1F9 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $2020.00
Specifications
| Antigen | IL-36 gamma/IL-1F9 |
|---|---|
| Clone | 278706 |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction 0.15-0.6 ug/mL |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2320SP
![]() |
R&D Systems
MAB2320SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2320100
![]() |
R&D Systems
MAB2320100 |
100 μg |
Each for $549.50
|
|
|||||
MAB2320500
![]() |
R&D Systems
MAB2320500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| IL-36 gamma/IL-1F9 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction 0.15-0.6 ug/mL | |
| Monoclonal | |
| Rat | |
| Human | |
| Q9NZH8 | |
| 56300 | |
| E. coli-derived recombinant human IL-36 gamma/IL-1F9 Met1-Asp169 Accession # Q9NZH8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-36 gamma/IL-1F9 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7, rhIL-36 beta/IL-1F8, rhIL-1ra/IL-1F3, or rhIL-18/IL-1F4 is observed. |
| 278706 | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1 epsilon, IL-1 H1, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, IL36 gamma, IL36G, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma | |
| IL36G | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title