Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-38/IL-1F10 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | IL-38/IL-1F10 |
---|---|
Clone | 316709 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blot.Specifications
IL-38/IL-1F10 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
Q8WWZ1 | |
84639 | |
E. coli-derived recombinant human IL-38/IL-1F10 Met1-Trp152 Accession # Q8WWZ1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-38/IL-1F10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse IL-1F10 is observed and no cross-reactivity with recombinant human (rh) IL-1 alpha/IL-1F1 rhIL-1 beta/IL-1F2, rhIL-1ra/IL-1F3, rhIL-18/IL-1F4, rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7/FIL1 zeta , rhIL-36 beta/IL-1F8, or rhIL-36 gamma/IL-1F9 is observed. |
316709 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
IL1F10 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title