Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6R alpha (Research Grade Sarilumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Supplier: R&D Systems MAB10346100
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
| IL-6R alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3570 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| Hu137 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Human IL-6R alpha | |
| 100 μg | |
| Primary | |
| Detects Human IL-6R alpha based on Sarilumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Sarilumab. This product is for research use only. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction