Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6R alpha (Research Grade Sarilumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | IL-6R alpha |
---|---|
Clone | Hu137 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10346SP
![]() |
R&D Systems
MAB10346SP |
25 μg |
Each for $214.00
|
|
|||||
MAB10346100
![]() |
R&D Systems
MAB10346100 |
100 μg |
Each for $514.50
|
|
|||||
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
IL-6R alpha | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
3570 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Hu137 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Human | |
Human | |
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
Human IL-6R alpha | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Human IL-6R alpha based on Sarilumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Sarilumab. This product is for research use only. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title